gRNA Design Details
- Target DNA Sequence
- GCACTACCCTGTCGGGATCC
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 100297493 - 100297515 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PKD2L1
- Entrez Gene ID
-
9033
- Gene Name
- polycystin 2 like 1, transient receptor potential cation channel
- Gene Aliases
-
PCL, PKD2L, PKDL, TRPP3
- Chromosome Location
-
Chr.10: 100288145 - 100346472 on GRCh38
- UniGene ID
- Hs.159241
- Species
-
Human