gRNA Design Details

Target DNA Sequence
CGAATCACCTTACAGAGTCT
PAM Sequence
GGG
Target locus
Chr.15: 69397487 - 69397509 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 90.72/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PAQR5
Entrez Gene ID
54852
Gene Name
progestin and adipoQ receptor family member 5
Gene Aliases
MPRG
Chromosome Location
Chr.15: 69284134 - 69407637 on GRCh38
UniGene ID
Hs.591096
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001104554.1 NP_001098024.1 7
NM_017705.3 NP_060175.3 7
XM_005254495.4 XP_005254552.1 5
XM_011521720.2 XP_011520022.1 8
XM_011521722.2 XP_011520024.1 8
XM_011521723.2 XP_011520025.1 6
XM_011521725.2 XP_011520027.1 5
XM_011521727.2 XP_011520029.1 7
XM_017022360.1 XP_016877849.1 10
XM_017022361.1 XP_016877850.1 8
XM_017022362.1 XP_016877851.1 9
XM_017022363.1 XP_016877852.1 6
XM_017022364.1 XP_016877853.1 7
XM_017022365.1 XP_016877854.1 8
XM_017022366.1 XP_016877855.1 6
XM_017022368.1 XP_016877857.1 4