gRNA Design Details
- Target DNA Sequence
- CTCCGGAGTGCATAAATCAG
- PAM Sequence
- AGG
- Target locus
-
Chr.18: 60371433 - 60371455 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MC4R
- Entrez Gene ID
-
4160
- Gene Name
- melanocortin 4 receptor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 60371331 - 60372768 on GRCh38
- UniGene ID
- Hs.532833
- Species
-
Human