gRNA Design Details
- Target DNA Sequence
- GGCGCTCCGCATGATCCTCG
- PAM Sequence
- CGG
- Target locus
-
Chr.7: 1092496 - 1092518 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
95.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPER1
- Entrez Gene ID
-
2852
- Gene Name
- G protein-coupled estrogen receptor 1
- Gene Aliases
-
CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
- Chromosome Location
-
Chr.7: 1086807 - 1093815 on GRCh38
- UniGene ID
- Hs.20961
- Species
-
Human