gRNA Design Details

Target DNA Sequence
GGCGCTCCGCATGATCCTCG
PAM Sequence
CGG
Target locus
Chr.7: 1092496 - 1092518 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 95.36/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GPER1
Entrez Gene ID
2852
Gene Name
G protein-coupled estrogen receptor 1
Gene Aliases
CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
Chromosome Location
Chr.7: 1086807 - 1093815 on GRCh38
UniGene ID
Hs.20961
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001039966.1 NP_001035055.1 3
NM_001098201.1 NP_001091671.1 2
NM_001505.2 NP_001496.1 2