gRNA Design Details
- Target DNA Sequence
- GAACAGCGCGGCGTAGAGCA
- PAM Sequence
- GGG
- Target locus
-
Chr.14: 52553308 - 52553330 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.28/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPR137C
- Entrez Gene ID
-
283554
- Gene Name
- G protein-coupled receptor 137C
- Gene Aliases
-
TM7SF1L2
- Chromosome Location
-
Chr.14: 52553148 - 52637713 on GRCh38
- UniGene ID
- Hs.416214
- Species
-
Human