gRNA Design Details
- Target DNA Sequence
- CATCGCCTACTACGACCACG
- PAM Sequence
- TGG
- Target locus
-
Chr.16: 89919795 - 89919817 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
94/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MC1R
- Entrez Gene ID
-
4157
- Gene Name
- melanocortin 1 receptor
- Gene Aliases
-
CMM5, MSH-R, SHEP2
- Chromosome Location
-
Chr.16: 89917879 - 89920977 on GRCh38
- UniGene ID
- Hs.513829
- Species
-
Human