gRNA Design Details
- Target DNA Sequence
- CCGTTTATTCCCTCGGGTAC
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 124746921 - 124746943 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
94.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPR37
- Entrez Gene ID
-
2861
- Gene Name
- G protein-coupled receptor 37
- Gene Aliases
-
EDNRBL, PAELR, hET(B)R-LP
- Chromosome Location
-
Chr.7: 124745601 - 124766025 on GRCh38
- UniGene ID
- Hs.406094
- Species
-
Human