gRNA Design Details
- Target DNA Sequence
- GAGGACTCACCAATCTGGTT
- PAM Sequence
- CGG
- Target locus
-
Chr.4: 22454868 - 22454890 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
79.2/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRA3
- Entrez Gene ID
-
166647
- Gene Name
- adhesion G protein-coupled receptor A3
- Gene Aliases
-
GPR125, PGR21, TEM5L
- Chromosome Location
-
Chr.4: 22387374 - 22516058 on GRCh38
- UniGene ID
- Hs.684429
- Species
-
Human