gRNA Design Details
- Target DNA Sequence
- CCGACAAAGGCATAGATGAT
- PAM Sequence
- GGG
- Target locus
-
Chr.3: 46373782 - 46373804 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.6/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CCR5
- Entrez Gene ID
-
1234
- Gene Name
- C-C motif chemokine receptor 5 (gene/pseudogene)
- Gene Aliases
-
CC-CKR-5, CCCKR5, CCR-5, CD195, CKR-5, CKR5, CMKBR5, IDDM22
- Chromosome Location
-
Chr.3: 46370142 - 46376206 on GRCh38
- UniGene ID
- Hs.450802
- Species
-
Human