gRNA Design Details
- Target DNA Sequence
- ACGTACAGCTGGTTGTCGCG
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 14162668 - 14162690 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
94.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRL1
- Entrez Gene ID
-
22859
- Gene Name
- adhesion G protein-coupled receptor L1
- Gene Aliases
-
CIRL1, CL1, LEC2, LPHN1
- Chromosome Location
-
Chr.19: 14147737 - 14206245 on GRCh38
- UniGene ID
- Hs.94229
- Species
-
Human