gRNA Design Details

Target DNA Sequence
AGCGTAAGCCAATACTGATG
PAM Sequence
AGG
Target locus
Chr.5: 68280590 - 68280612 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 96.18/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PIK3R1
Entrez Gene ID
5295
Gene Name
phosphoinositide-3-kinase regulatory subunit 1
Gene Aliases
AGM7, GRB1, IMD36, p85, p85-ALPHA
Chromosome Location
Chr.5: 68215737 - 68301821 on GRCh38
UniGene ID
Hs.132225
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_181523.2 NP_852664.1 6
XM_005248542.3 XP_005248599.1 6
XM_011543493.2 XP_011541795.1 5
XM_017009585.1 XP_016865074.1 6
XM_017009586.1 XP_016865075.1 5