gRNA Design Details
- Target DNA Sequence
- CGATCGAGAGGTAGCGCTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.14: 52314941 - 52314963 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PTGER2
- Entrez Gene ID
-
5732
- Gene Name
- prostaglandin E receptor 2
- Gene Aliases
-
EP2
- Chromosome Location
-
Chr.14: 52314298 - 52328606 on GRCh38
- UniGene ID
- Hs.2090
- Species
-
Human