gRNA Design Details
- Target DNA Sequence
- TTCGGGACTTCATCATCCTA
- PAM Sequence
- CGG
- Target locus
-
Chr.6: 47714763 - 47714785 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.46/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRF4
- Entrez Gene ID
-
221393
- Gene Name
- adhesion G protein-coupled receptor F4
- Gene Aliases
-
GPR115, PGR18
- Chromosome Location
-
Chr.6: 47698553 - 47722021 on GRCh38
- UniGene ID
- Hs.150131
- Species
-
Human