gRNA Design Details
- Target DNA Sequence
- TGTAGCGGAAGTCTCGGTTC
- PAM Sequence
- CGG
- Target locus
-
Chr.17: 15975219 - 15975241 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
95.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADORA2B
- Entrez Gene ID
-
136
- Gene Name
- adenosine A2b receptor
- Gene Aliases
-
ADORA2
- Chromosome Location
-
Chr.17: 15927781 - 15975896 on GRCh38
- UniGene ID
- Hs.167046
- Species
-
Human