gRNA Design Details
- Target DNA Sequence
- GAACAACACTATCTCAGCCA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 78938126 - 78938148 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRL4
- Entrez Gene ID
-
64123
- Gene Name
- adhesion G protein-coupled receptor L4
- Gene Aliases
-
ELTD1, ETL, KPG_003
- Chromosome Location
-
Chr.1: 78889764 - 79006810 on GRCh38
- UniGene ID
- Hs.132314
- Species
-
Human