gRNA Design Details

Target DNA Sequence
TGTTCGGTTACTTGTGGTCA
PAM Sequence
AGG
Target locus
Chr.6: 68936557 - 68936579 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 95.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRB3
Entrez Gene ID
577
Gene Name
adhesion G protein-coupled receptor B3
Gene Aliases
BAI3
Chromosome Location
Chr.6: 68635282 - 69389511 on GRCh38
UniGene ID
Hs.13261
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq XM_005248752.2 XP_005248809.1 4