gRNA Design Details
- Target DNA Sequence
- ACAACCACTAGAGTCCTGTA
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 109119099 - 109119121 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TRHR
- Entrez Gene ID
-
7201
- Gene Name
- thyrotropin releasing hormone receptor
- Gene Aliases
-
TRH-R
- Chromosome Location
-
Chr.8: 109085745 - 109119785 on GRCh38
- UniGene ID
- Hs.3022
- Species
-
Human