gRNA Design Details

Target DNA Sequence
ACCCTGCGAGTGCCATCAGG
PAM Sequence
AGG
Target locus
Chr.14: 80955761 - 80955783 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 87.8/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TSHR
Entrez Gene ID
7253
Gene Name
thyroid stimulating hormone receptor
Gene Aliases
CHNG1, LGR3, hTSHR-I
Chromosome Location
Chr.14: 80954989 - 81146302 on GRCh38
UniGene ID
Hs.160411
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000369.2 NP_000360.2 1
NM_001018036.2 NP_001018046.1 1
NM_001142626.2 NP_001136098.1 1
XM_005268037.4 XP_005268094.1 2
XM_005268039.1 XP_005268096.1 2
XM_006720245.1 XP_006720308.1 2
XM_011537119.2 XP_011535421.1 1