gRNA Design Details
- Target DNA Sequence
- GAAGCGTATGAGGTGAAGCT
- PAM Sequence
- GGG
- Target locus
-
Chr.7: 127927235 - 127927257 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.35/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss8
- Entrez Gene ID
-
76560
- Gene Name
- protease, serine 8 (prostasin)
- Gene Aliases
-
2410039E18Rik, AI313909, C79772, CAP1, fr, mCAP1
- Chromosome Location
-
Chr.7: 127925717 - 127930142 on GRCm38
- UniGene ID
- Mm.5875
- Species
-
Mouse