gRNA Design Details
- Target DNA Sequence
- ACTGGCGACGATGATGCTCT
- PAM Sequence
- CGG
- Target locus
-
Chr.5: 116591360 - 116591382 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.23/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Srrm4
- Entrez Gene ID
-
68955
- Gene Name
- serine/arginine repetitive matrix 4
- Gene Aliases
-
1500001A10Rik, B230202K19Rik, bv, mKIAA1853, nSR100
- Chromosome Location
-
Chr.5: 116439272 - 116592562 on GRCm38
- UniGene ID
- Mm.146779
- Species
-
Mouse