gRNA Design Details
- Target DNA Sequence
- TTCAAGGGCCATTGCTGCCA
- PAM Sequence
- CGG
- Target locus
-
Chr.4: 21854379 - 21854401 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.34/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pnisr
- Entrez Gene ID
-
66625
- Gene Name
- PNN interacting serine/arginine-rich
- Gene Aliases
-
5730406M06Rik, AA673174, Pinsr, Sfrs18, Srsf18
- Chromosome Location
-
Chr.4: 21847583 - 21876475 on GRCm38
- UniGene ID
- Mm.100117
- Species
-
Mouse