gRNA Design Details
- Target DNA Sequence
- TATGGTAGGGTGGCTCGTGT
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 102863670 - 102863692 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sgip1
- Entrez Gene ID
-
73094
- Gene Name
- SH3-domain GRB2-like (endophilin) interacting protein 1
- Gene Aliases
-
3110007P09Rik, SGIP1alpha, gag
- Chromosome Location
-
Chr.4: 102759167 - 102972701 on GRCm38
- UniGene ID
- Mm.238094
- Species
-
Mouse