gRNA Design Details
- Target DNA Sequence
- GTGCGAAGTGCACAATGGTG
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 151439311 - 151439333 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.49/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrl4
- Entrez Gene ID
-
170757
- Gene Name
- adhesion G protein-coupled receptor L4
- Gene Aliases
-
1110033N21Rik, ETL1, Eltd1, Etl
- Chromosome Location
-
Chr.3: 151437882 - 151545081 on GRCm38
- UniGene ID
- Mm.317261
- Species
-
Mouse