gRNA Design Details

Target DNA Sequence
TCGGTGATCTTTGGTCTGAA
PAM Sequence
AGG
Target locus
Chr.1: 91387314 - 91387336 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 85.22/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Ilkap
Entrez Gene ID
67444
Gene Name
integrin-linked kinase-associated serine/threonine phosphatase 2C
Gene Aliases
0710007A14Rik, 1600009O09Rik, PP2C-DELTA
Chromosome Location
Chr.1: 91373846 - 91398841 on GRCm38
UniGene ID
Mm.337240
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_023343.2 NP_075832.1 5