gRNA Design Details
- Target DNA Sequence
- TGCCACATCAGGTTCCCAGG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 91388435 - 91388457 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.35/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ilkap
- Entrez Gene ID
-
67444
- Gene Name
- integrin-linked kinase-associated serine/threonine phosphatase 2C
- Gene Aliases
-
0710007A14Rik, 1600009O09Rik, PP2C-DELTA
- Chromosome Location
-
Chr.1: 91373846 - 91398841 on GRCm38
- UniGene ID
- Mm.337240
- Species
-
Mouse