gRNA Design Details
- Target DNA Sequence
- TTCAATCGTGCCCAGAGATC
- PAM Sequence
- CGG
- Target locus
-
Chr.6: 83054176 - 83054198 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.49/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Htra2
- Entrez Gene ID
-
64704
- Gene Name
- HtrA serine peptidase 2
- Gene Aliases
-
AI481710, Omi, Prss25, mnd2
- Chromosome Location
-
Chr.6: 83051266 - 83054571 on GRCm38
- UniGene ID
- Mm.21880
- Species
-
Mouse