gRNA Design Details
- Target DNA Sequence
- TGTGTCCTCGACCGCTTCAC
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 126128826 - 126128848 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.6/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk40
- Entrez Gene ID
-
74178
- Gene Name
- serine/threonine kinase 40
- Gene Aliases
-
2310004N11Rik, AI428792
- Chromosome Location
-
Chr.4: 126103874 - 126141029 on GRCm38
- UniGene ID
- Mm.440269
- Species
-
Mouse