gRNA Design Details
- Target DNA Sequence
- ACTGTGCACACGCAAGTGGC
- PAM Sequence
- TGG
- Target locus
-
Chr.16: 33710525 - 33710547 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
83.11/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Heg1
- Entrez Gene ID
-
77446
- Gene Name
- HEG homolog 1 (zebrafish)
- Gene Aliases
-
4632417D23Rik, 5530401I02Rik, 9530025L16Rik, BC038614, Gm629, OTTMUSG00000027315
- Chromosome Location
-
Chr.16: 33684430 - 33771576 on GRCm38
- UniGene ID
- Mm.245741
- Species
-
Mouse