gRNA Design Details
- Target DNA Sequence
- TGCATTGCCAATTAAGCTCA
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 42669827 - 42669849 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.1/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrf4
- Entrez Gene ID
-
78249
- Gene Name
- adhesion G protein-coupled receptor F4
- Gene Aliases
-
4632435A09Rik, AV239010, Gpr115
- Chromosome Location
-
Chr.17: 42656887 - 42692284 on GRCm38
- UniGene ID
- Mm.66217
- Species
-
Mouse