gRNA Design Details

Target DNA Sequence
TCCTGCAGCTAGCCTATCCT
PAM Sequence
TGG
Target locus
Chr.17: 42669868 - 42669890 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 84.62/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Adgrf4
Entrez Gene ID
78249
Gene Name
adhesion G protein-coupled receptor F4
Gene Aliases
4632435A09Rik, AV239010, Gpr115
Chromosome Location
Chr.17: 42656887 - 42692284 on GRCm38
UniGene ID
Mm.66217
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001289499.1 NP_001276428.1 5
NM_001289500.1 NP_001276429.1 5
NM_001289501.1 NP_001276430.1 5
NM_030067.2 NP_084343.1 5