gRNA Design Details

Target DNA Sequence
CTTGGATGAGAAGGTGACGA
PAM Sequence
TGG
Target locus
Chr.4: 58205984 - 58206006 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 85.09/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Svep1
Entrez Gene ID
64817
Gene Name
sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
Gene Aliases
1110021D17Rik, 4833413O10Rik, AA387071, D430029O09Rik, Polydom
Chromosome Location
Chr.4: 58042796 - 58206596 on GRCm38
UniGene ID
Mm.261024
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_022814.2 NP_073725.2 1