gRNA Design Details
- Target DNA Sequence
- GTACGCAGATCCGGGATTTC
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 34037719 - 34037741 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Bphl
- Entrez Gene ID
-
68021
- Gene Name
- biphenyl hydrolase-like (serine hydrolase, breast epithelial mucin-associated antigen)
- Gene Aliases
-
2010012D11Rik, 5730533B08Rik, AI115341
- Chromosome Location
-
Chr.13: 34037593 - 34074074 on GRCm38
- UniGene ID
- Mm.140243
- Species
-
Mouse