gRNA Design Details

Target DNA Sequence
AAAATCTCGATCTGGGGGTC
PAM Sequence
TGG
Target locus
Chr.13: 34046879 - 34046901 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 88.29/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Bphl
Entrez Gene ID
68021
Gene Name
biphenyl hydrolase-like (serine hydrolase, breast epithelial mucin-associated antigen)
Gene Aliases
2010012D11Rik, 5730533B08Rik, AI115341
Chromosome Location
Chr.13: 34037593 - 34074074 on GRCm38
UniGene ID
Mm.140243
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_026512.1 NP_080788.1 3