gRNA Design Details
- Target DNA Sequence
- GACCTGACGAACAACCGAAT
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 50013441 - 50013463 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
93.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgra2
- Entrez Gene ID
-
70693
- Gene Name
- adhesion G protein-coupled receptor A2
- Gene Aliases
-
3830613O22Rik, AU044632, Gpr125, Tem5-like
- Chromosome Location
-
Chr.5: 49959951 - 50058996 on GRCm38
- UniGene ID
- Mm.272974
- Species
-
Mouse