gRNA Design Details
- Target DNA Sequence
- CAAAACAGTGCAAGCGTTCA
- PAM Sequence
- TGG
- Target locus
-
Chr.15: 99173485 - 99173507 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.1/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spats2
- Entrez Gene ID
-
72572
- Gene Name
- spermatogenesis associated, serine-rich 2
- Gene Aliases
-
2700012F11Rik, 59kDa, Scr59, p59
- Chromosome Location
-
Chr.15: 99125926 - 99213215 on GRCm38
- UniGene ID
- Mm.276650
- Species
-
Mouse