gRNA Design Details
- Target DNA Sequence
- GTCCAATACTGTCCTGGCCC
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 99167108 - 99167130 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.52/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spats2
- Entrez Gene ID
-
72572
- Gene Name
- spermatogenesis associated, serine-rich 2
- Gene Aliases
-
2700012F11Rik, 59kDa, Scr59, p59
- Chromosome Location
-
Chr.15: 99125926 - 99213215 on GRCm38
- UniGene ID
- Mm.276650
- Species
-
Mouse