gRNA Design Details
- Target DNA Sequence
- GCAAGAGCACTTGTAGGAAG
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 109864142 - 109864164 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.99/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Scube2
- Entrez Gene ID
-
56788
- Gene Name
- signal peptide, CUB domain, EGF-like 2
- Gene Aliases
-
4932442O19Rik, Cegf1, Cegp1
- Chromosome Location
-
Chr.7: 109798690 - 109865679 on GRCm38
- UniGene ID
- Mm.40447
- Species
-
Mouse