gRNA Design Details
- Target DNA Sequence
- CCGGACATTAGTGGTGGCTA
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 40516153 - 40516175 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss37
- Entrez Gene ID
-
67690
- Gene Name
- protease, serine 37
- Gene Aliases
-
1700016G05Rik, Tryx2
- Chromosome Location
-
Chr.6: 40514820 - 40519542 on GRCm38
- UniGene ID
- Mm.52550
- Species
-
Mouse