gRNA Design Details

Target DNA Sequence
ACCGACAACAGGCTCAAGAG
PAM Sequence
TGG
Target locus
Chr.9: 63617514 - 63617536 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 87.61/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Aagab
Entrez Gene ID
66939
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
2310007F21Rik, p34
Chromosome Location
Chr.9: 63602629 - 63641897 on GRCm38
UniGene ID
Mm.34338
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_025857.2 NP_080133.1 4