gRNA Design Details
- Target DNA Sequence
- CCGAATGCACCACGGCTAAT
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 23155725 - 23155747 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mastl
- Entrez Gene ID
-
67121
- Gene Name
- microtubule associated serine/threonine kinase-like
- Gene Aliases
-
2700091H24Rik, C88295, GW, GWL, MAST-L, THC2
- Chromosome Location
-
Chr.2: 23116544 - 23156024 on GRCm38
- UniGene ID
- Mm.253721
- Species
-
Mouse