gRNA Design Details
- Target DNA Sequence
- CTTCGACGCCAAAGCGAAAT
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 89510685 - 89510707 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss23
- Entrez Gene ID
-
76453
- Gene Name
- protease, serine 23
- Gene Aliases
-
2310046G15Rik, 4930479H08Rik, AI790574, Spuve
- Chromosome Location
-
Chr.7: 89507785 - 89527254 on GRCm38
- UniGene ID
- Mm.250438
- Species
-
Mouse