gRNA Design Details
- Target DNA Sequence
- GCTGTGAGTAGATGCCACGG
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 75525451 - 75525473 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk11ip
- Entrez Gene ID
-
71728
- Gene Name
- serine/threonine kinase 11 interacting protein
- Gene Aliases
-
1200014D22Rik, BB131189, LIP1, LKB1IP
- Chromosome Location
-
Chr.1: 75521529 - 75537335 on GRCm38
- UniGene ID
- Mm.312650
- Species
-
Mouse