gRNA Design Details
- Target DNA Sequence
- GAGTTCCCCAGGACTTACGG
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 115330562 - 115330584 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Srsf9
- Entrez Gene ID
-
108014
- Gene Name
- serine/arginine-rich splicing factor 9
- Gene Aliases
-
25kDa, 2610029M16Rik, SRp30c, Sfrs9
- Chromosome Location
-
Chr.5: 115327161 - 115333084 on GRCm38
- UniGene ID
- Mm.287826
- Species
-
Mouse