gRNA Design Details
- Target DNA Sequence
- CACCTGCCAATATTAAGAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 30519688 - 30519710 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.61/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ssmem1
- Entrez Gene ID
-
75647
- Gene Name
- serine-rich single-pass membrane protein 1
- Gene Aliases
-
1700016K02Rik, 1700025E21Rik
- Chromosome Location
-
Chr.6: 30509849 - 30520253 on GRCm38
- UniGene ID
- Mm.46117
- Species
-
Mouse