gRNA Design Details
- Target DNA Sequence
- GCTCAACAACAACCCGAGGA
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 105157887 - 105157909 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.03/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ppap2b
- Entrez Gene ID
-
67916
- Gene Name
- phosphatidic acid phosphatase type 2B
- Gene Aliases
-
1110003O22Rik, 2610002D05Rik, AV025606, D4Bwg0538e, D4Bwg1535e, Lpp3, Ppab2b
- Chromosome Location
-
Chr.4: 105157347 - 105232767 on GRCm38
- UniGene ID
- Mm.348326
- Species
-
Mouse