gRNA Design Details
- Target DNA Sequence
- TGAGGTGCTCTGAAGAACGG
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 112853133 - 112853155 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gpr132
- Entrez Gene ID
-
56696
- Gene Name
- G protein-coupled receptor 132
- Gene Aliases
-
G2a
- Chromosome Location
-
Chr.12: 112850873 - 112868418 on GRCm38
- UniGene ID
- Mm.20455
- Species
-
Mouse