gRNA Design Details
- Target DNA Sequence
- TGACAACTACCAAGGAGATT
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 119370194 - 119370216 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.61/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adipor2
- Entrez Gene ID
-
68465
- Gene Name
- adiponectin receptor 2
- Gene Aliases
-
1110001I14Rik, ADCR2, AI115388, AW554121, D6Ucla1e, Paqr2
- Chromosome Location
-
Chr.6: 119353150 - 119417483 on GRCm38
- UniGene ID
- Mm.291826
- Species
-
Mouse