gRNA Design Details
- Target DNA Sequence
- GAAGAGGAGGAATGATCTCC
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 70534516 - 70534538 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
70.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Megf9
- Entrez Gene ID
-
230316
- Gene Name
- multiple EGF-like-domains 9
- Gene Aliases
-
4933405H16Rik, 9630025C07, Egfl5, mKIAA0818
- Chromosome Location
-
Chr.4: 70431927 - 70534928 on GRCm38
- UniGene ID
- Mm.251188
- Species
-
Mouse