gRNA Design Details
- Target DNA Sequence
- GATGCCAACTACAACCTGCT
- PAM Sequence
- GGG
- Target locus
-
Chr.19: 42334028 - 42334050 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
71.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MEGF8
- Entrez Gene ID
-
1954
- Gene Name
- multiple EGF like domains 8
- Gene Aliases
-
C19orf49, CRPT2, EGFL4, SBP1
- Chromosome Location
-
Chr.19: 42325609 - 42378769 on GRCh38
- UniGene ID
- Hs.132483
- Species
-
Human